siRNA Details
Gene Symbol: Pla2g4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_230487.7 | XP_230487.5 | 13 | 2004 |

More Information
Other Species Targeted

Panther Classification:

Gene Symbol: Pla2g4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_230487.7 | XP_230487.5 | 13 | 2004 |