Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106503.1 | NP_001099973.1 | 1, 2 | 652 | ||
XM_006234917.2 | XP_006234979.1 | 3, 4 | 910 | |||
XM_008762156.1 | XP_008760378.1 | 3, 4 | 914 |