Search
Search

Ing4| Gene Symbol: Ing4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079887.1 | NP_001073356.1 | 5 | 468 | ||
| XM_006237349.2 | XP_006237411.1 | 5 | 579 | |||
| XM_006237350.2 | XP_006237412.1 | 5 | 576 | |||
| GenBank mRNA | BC086570.1 | - | 5 | 468 | ||
| BC097942.1 | - | 4 | 651 | |||
| BC127511.1 | - | 5 | 468 | 7376723 | ||
| FQ213328.1 | - | 2 | 105 | |||
| FQ232370.1 | - | 1 | 102 | |||