Search
Search

Mroh7| Gene Symbol: Mroh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100965.2 | NP_001094435.1 | 2 | 912 | ||
| XM_008763860.1 | XP_008762082.1 | 4 | 1171 | |||
| XM_008763861.1 | XP_008762083.1 | 3 | 1100 | |||
| XM_008763862.1 | XP_008762084.1 | 4 | 1072 | |||
| XM_008763863.1 | XP_008762085.1 | 4 | 1454 | |||
| XM_008763864.1 | XP_008762086.1 | 3 | 1001 | |||
| XM_008763865.1 | XP_008762087.1 | 3 | 929 | |||
| XM_008763866.1 | XP_008762088.1 | 4 | 1243 | |||
| GenBank mRNA | BC133066.1 | - | 2 | 915 | 7134332 | |