Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135709.1 | NP_001129181.1 | 14 | 2257 | ||
XM_006238858.2 | XP_006238920.1 | 12 | 2249 |