Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039325.1 | NP_001034414.1 | 2, 3 | 709 | ||
GenBank mRNA | AY886537.1 | - | 2, 3 | 709 |