siRNA Details
Gene Symbol: RGD1304745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766002.1 | XP_008764224.1 | 1 | 552 |

Gene Symbol: RGD1304745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766002.1 | XP_008764224.1 | 1 | 552 |