Search
Search

Aph1b
LOC100911293
LOC691658| Gene Symbol: Aph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047090.1 | NP_001040555.1 | 3 | 454 | ||
| GenBank mRNA | BC098711.1 | - | 3 | 316 | ||
| BC166592.1 | - | 3 | 410 | 7929867 | ||
| DQ672552.1 | - | 3 | 454 | |||
| Gene Symbol: LOC100911293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766401.1 | XP_008764623.1 | 449 | |||
| Gene Symbol: LOC691658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001079175.5 | XP_001079175.2 | 3 | 484 | ||
| XM_003750532.3 | XP_003750580.1 | 3 | 483 | |||
| XM_006243318.2 | XP_006243380.1 | 3 | 691 | |||
| XM_006243319.2 | XP_006243381.1 | 3 | 695 | |||