siRNA Details
Gene Symbol: Sox14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106850.1 | NP_001100320.1 | 1 | 980 |

More Information
Other Species Targeted

Panther Classification:

Gene Symbol: Sox14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106850.1 | NP_001100320.1 | 1 | 980 |