siRNA Details
Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_217313.8 | XP_217313.6 | 4 | 2230 | ||
GenBank mRNA | FQ231323.1 | - | 4 | 517 |

More Information
Other Species Targeted


Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_217313.8 | XP_217313.6 | 4 | 2230 | ||
GenBank mRNA | FQ231323.1 | - | 4 | 517 |