Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106898.1 | NP_001100368.1 | 2 | 614 |
Set Membership: |
G-protein coupled receptor |