Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLMAP SIRNA Assays ›
Gene Symbol:
SLMAP
Gene Name:
sarcolemma associated protein
Gene Aliases:
SLAP
Chromosome Location:
Chr.3: 57756221 - 57930013 on Build GRCh38
UniGene ID:
Hs.476432
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15433
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLMAP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001304420.2
NP_001291349.1
13
1673
NM_001304421.2
NP_001291350.1
11
1559
NM_001304422.2
NP_001291351.1
1
155
NM_001304423.2
NP_001291352.1
1
155
NM_001311178.1
NP_001298107.1
1
155
NM_001311179.1
NP_001298108.1
1
155
NM_007159.4
NP_009090.2
12
1622
XM_005265456.3
XP_005265513.1
15
2560
XM_005265457.3
XP_005265514.1
15
2554
XM_005265458.3
XP_005265515.1
15
2560
XM_005265459.3
XP_005265516.1
14
2509
XM_005265460.3
XP_005265517.1
14
2509
XM_005265461.3
XP_005265518.1
14
2503
XM_005265462.3
XP_005265519.1
14
2497
XM_005265464.3
XP_005265521.1
14
2491
XM_005265465.3
XP_005265522.1
13
2458
XM_005265466.3
XP_005265523.1
13
2452
XM_005265467.3
XP_005265524.1
13
2446
XM_005265468.3
XP_005265525.1
15
2560
XM_005265470.3
XP_005265527.1
12
2395
XM_005265471.3
XP_005265528.1
12
2389
XM_005265472.3
XP_005265529.1
13
2446
XM_006713326.2
XP_006713389.1
13
2452
XM_011534103.1
XP_011532405.1
13
2446
XM_011534104.1
XP_011532406.1
13
2458
XM_011534105.1
XP_011532407.1
12
2395
XM_017007146.1
XP_016862635.1
15
2554
XM_017007147.1
XP_016862636.1
14
2503
XM_017007148.1
XP_016862637.1
14
2509
XM_017007149.1
XP_016862638.1
14
2503
XM_017007150.1
XP_016862639.1
14
2503
XM_017007151.1
XP_016862640.1
13
2458
XM_017007152.1
XP_016862641.1
13
2452
XM_017007153.1
XP_016862642.1
13
2440
XM_017007154.1
XP_016862643.1
13
2440
XM_017007155.1
XP_016862644.1
15
2554
XM_017007156.1
XP_016862645.1
15
2560
XM_017007157.1
XP_016862646.1
15
2554
XM_017007158.1
XP_016862647.1
12
2395
XM_017007159.1
XP_016862648.1
14
2509
XM_017007160.1
XP_016862649.1
12
2389
XM_017007161.1
XP_016862650.1
14
2503
XM_017007162.1
XP_016862651.1
14
2509
XM_017007163.1
XP_016862652.1
14
2503
XM_017007164.1
XP_016862653.1
13
2452
XM_017007165.1
XP_016862654.1
13
2440
XM_017007166.1
XP_016862655.1
13
2440
XM_017007167.1
XP_016862656.1
12
2395
XM_017007168.1
XP_016862657.1
12
2389
XM_017007169.1
XP_016862658.1
15
2560
GenBank mRNA
AB046821.1
-
10
863
AF304450.1
-
1
28
AK124200.1
-
13
1363
AK300106.1
-
1
78
AK304493.1
-
1
82
AY358410.1
-
12
1560
BC114627.1
-
11
1257
40081892
BC115701.1
-
13
1371
40081893
CR627321.1
-
1
12
DB100357.1
-
1
159
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0