Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013965.1 | NP_001013987.1 | 2, 3 | 728 | ||
GenBank mRNA | AB194013.1 | - | 2, 3 | 613 | ||
BC079304.1 | - | 2, 3 | 728 | 7134598 |