Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014234.4 | NP_055049.1 | 6 | 639 | ||
GenBank mRNA | BC008185.1 | - | 6 | 634 | 4213813 | |
BT007239.1 | - | 6 | 607 | |||
D82061.1 | - | 6 | 603 |