Search
Search

Cntrob| Gene Symbol: Cntrob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134645.1 | NP_001128117.1 | 16 | 2526 | ||
| XM_006246701.2 | XP_006246763.1 | 17 | 2900 | |||
| XM_006246702.2 | XP_006246764.1 | 15 | 3110 | |||
| XM_008767825.1 | XP_008766047.1 | 16 | 2234 | |||
| GenBank mRNA | EF532342.1 | - | 16 | 2526 | ||
| EF532343.1 | - | 16 | 2578 | |||
| EF532344.1 | - | 16 | 2585 | |||
| EF532345.1 | - | 16 | 2578 | |||
| EF532346.1 | - | 15 | 2848 | |||
| EF532347.1 | - | 16 | 2896 | |||
| EF532348.1 | - | 17 | 2922 | |||
| EF532349.1 | - | 17 | 2954 | |||
| EF532350.1 | - | 15 | 2932 | |||