Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134534.1 | NP_001128006.1 | 3 | 1807 | ||
XM_006247691.2 | XP_006247753.1 | 3 | 1832 | |||
XM_006247692.2 | XP_006247754.1 | 3 | 2603 | |||
XM_008768371.1 | XP_008766593.1 | 2 | 2262 |