Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013980.1 | NP_001014002.1 | 2 | 404 | ||
GenBank mRNA | BC087052.1 | - | 2 | 404 | 7122930 |