Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037191.1 | NP_001032268.1 | 10 | 1371 | ||
GenBank mRNA | BC103478.1 | - | 10 | 1371 | 7937275 |