Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107138.1 | NP_001100608.1 | 16 | 2094 | ||
XM_006249300.2 | XP_006249362.1 | 16 | 2671 | |||
XM_006249301.2 | XP_006249363.1 | 16 | 2668 | |||
XM_008769224.1 | XP_008767446.1 | 15 | 2566 | |||
XM_008769225.1 | XP_008767447.1 | 15 | 2563 | |||
XM_008769226.1 | XP_008767448.1 | 13 | 1576 | |||
GenBank mRNA | FQ230302.1 | - | 7 | 783 | ||
FQ232681.1 | - | 7 | 783 | |||
FQ233202.1 | - | 6 | 682 |