Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767478.1 | XP_008765700.1 | 11 | 3023 |
Gene Symbol: LOC103693257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767457.1 | XP_008765679.1 | 11 | 3004 |