Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107169.1 | NP_001100639.1 | 10 | 1733 | ||
XM_008769455.1 | XP_008767677.1 | 11 | 2354 |