Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107231.2 | NP_001100701.1 | 1 | 95 | ||
NM_001277214.1 | NP_001264143.1 | 1 | 95 | |||
NM_001277215.1 | NP_001264144.1 | 1 | 95 | |||
XM_006251292.2 | XP_006251354.1 | 1 | 117 |