Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004819.2 | NP_004810.2 | 3 | 405 | ||
XM_005259286.1 | XP_005259343.1 | 3 | 358 | |||
XM_011527354.1 | XP_011525656.1 | 4 | 408 | |||
GenBank mRNA | AB209024.1 | - | 3 | 2653 | ||
AB210010.1 | - | 3 | 250 | |||
AF049526.1 | - | 3 | 195 | |||
AK130602.1 | - | 3 | 262 | |||
AK300252.1 | - | 3 | 259 | |||
BC006536.2 | - | 2 | 112 | |||
BC006567.2 | - | 2 | 112 | |||
BC030214.1 | - | 3 | 247 | |||
BX649165.1 | - | 3 | 260 | |||
DB044963.1 | - | 3 | 381 | |||
DB461873.1 | - | 3 | 405 | |||
U88726.1 | - | 2 | 176 | |||
Y10931.1 | - | 3 | 224 |