Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thtpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007682.1 | NP_001007683.1 | 1 | 487 | ||
XM_006251970.2 | XP_006252032.1 | 1 | 2418 | |||
GenBank mRNA | BC081978.1 | - | 1 | 487 | 7128058 |