Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012041.1 | NP_001012041.1 | 9 | 1132 | ||
XM_006251974.2 | XP_006252036.1 | 9 | 1260 | |||
XM_006251975.1 | XP_006252037.1 | 9 | 1204 | |||
XM_008770679.1 | XP_008768901.1 | 9 | 1245 | |||
GenBank mRNA | BC079454.1 | - | 9 | 1132 | 7121327 |