Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204466.1 | NP_001191395.1 | 12 | 1469 | ||
NM_001204467.1 | NP_001191396.1 | 13 | 1697 | |||
NM_001204468.1 | NP_001191397.1 | 13 | 1731 | |||
NM_005676.4 | NP_005667.2 | 13 | 1700 | |||
NM_152856.2 | NP_690595.1 | 12 | 1466 | |||
XM_005272677.4 | XP_005272734.1 | 13 | 1750 | |||
XM_005272678.4 | XP_005272735.1 | 12 | 1284 | |||
XM_005272679.4 | XP_005272736.1 | 12 | 1281 | |||
XM_006724563.1 | XP_006724626.1 | 9 | 917 | |||
XM_011543989.2 | XP_011542291.1 | 5 | 641 | |||
XM_017029884.1 | XP_016885373.1 | 12 | 1620 | |||
XM_017029885.1 | XP_016885374.1 | 12 | 1392 | |||
GenBank mRNA | AK025854.1 | - | 1 | 73 | ||
AK292758.1 | - | 12 | 1420 | |||
AL137421.1 | - | 14 | 1589 | |||
BC000681.2 | - | 10 | 922 | |||
BC003089.1 | - | 12 | 1405 | 3536058 | ||
BC004181.2 | - | 13 | 1653 | 2961094 | ||
BC008733.2 | - | 13 | 1685 | 3163064 | ||
BC024153.2 | - | 12 | 1315 | 3845258 | ||
BX537969.1 | - | 13 | 1682 | |||
D50912.1 | - | 13 | 1548 | |||
KU178411.1 | - | 11 | 1073 | |||
KU178412.1 | - | 12 | 1307 |