Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271295.1 | NP_001258224.1 | 2 | 2055 | ||
NM_001271296.1 | NP_001258225.1 | 2 | 2055 |
Gene Symbol: LOC103694119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771839.1 | XP_008770061.1 | 2 | 733 |