Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254139.2 | XP_006254201.1 | 13 | 2270 | ||
XM_008771806.1 | XP_008770028.1 | 12 | 1792 | |||
XM_008771807.1 | XP_008770029.1 | 12 | 1723 | |||
XM_008771808.1 | XP_008770030.1 | 12 | 1792 | |||
XM_008771809.1 | XP_008770031.1 | 12 | 1792 | |||
XM_008771810.1 | XP_008770032.1 | 12 | 1792 |