Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007690.1 | NP_001007691.1 | 2 | 1218 | ||
XM_006227640.2 | XP_006227702.1 | 2 | 1260 | |||
GenBank mRNA | BC083823.1 | - | 2 | 1218 | 7111792 |