Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107512.1 | NP_001100982.1 | 3 | 399 | ||
XM_006229069.1 | XP_006229131.1 | 3 | 354 | |||
XM_006229070.2 | XP_006229132.1 | 3 | 356 | |||
XM_008759378.1 | XP_008757600.1 | 3 | 438 | |||
GenBank mRNA | BC168947.1 | - | 3 | 348 | 9027778 |