Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199101.1 | NP_954532.1 | 17 | 2070 | ||
XM_006229071.1 | XP_006229133.1 | 16 | 2010 | |||
XM_006229072.1 | XP_006229134.1 | 17 | 2226 | |||
GenBank mRNA | BC061738.1 | - | 17 | 2070 | 5599064 |