Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005885.2 | NP_001005885.1 | 2 | 104 | ||
XM_006229766.2 | XP_006229828.1 | 2 | 172 | |||
GenBank mRNA | AY325765.1 | - | 2 | 144 | ||
AY380804.1 | - | 2 | 162 | |||
BC098002.1 | - | 2 | 104 | 7453525 |