Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104640.1 | NP_001098110.1 | 5 | 659 | ||
XM_006256574.2 | XP_006256636.1 | 5 | 726 | |||
GenBank mRNA | DQ503410.1 | - | 5 | 659 |