Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271167.1 | NP_001258096.1 | 11 | 1339 | ||
XM_008761099.1 | XP_008759321.1 | 12 | 2824 | |||
XM_008761100.1 | XP_008759322.1 | 12 | 1456 | |||
XM_008761101.1 | XP_008759323.1 | 11 | 2734 |