Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107701.1 | NP_001101171.1 | 3, 4 | 848 | ||
XM_006232671.2 | XP_006232733.1 | 3, 4 | 453 | |||
GenBank mRNA | BC087156.1 | - | 3, 4 | 484 |