Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749357.3 | XP_003749405.1 | 8 | 4281 | ||
XM_006233013.2 | XP_006233075.1 | 8 | 3235 | |||
XM_006233014.2 | XP_006233076.1 | 8 | 3235 | |||
XM_006233015.2 | XP_006233077.1 | 8 | 3013 |