Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NGF SIRNA Assays ›
          Gene Symbol:
          Ngf
          Gene Name:
          nerve growth factor (beta polypeptide)
          Gene Aliases:
          Ngfb, beta-NGF
          Chromosome Location:
          Chr.2: 204886158 - 204939523 on Build Rnor_6.0
          UniGene ID:
          Rn.22168
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s160631

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ngf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277055.1 NP_001263984.1 3 483
          XM_006233052.2 XP_006233114.1 3 579
          XM_006233053.2 XP_006233115.1 3 514
          GenBank mRNA FQ232560.1 - 3 483

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          neurotrophic factor

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          cell-cell signaling
          peripheral nervous system development
          aging
          memory
          circadian rhythm
          positive regulation of cell proliferation
          adult locomotory behavior
          extrinsic apoptotic signaling pathway via death domain receptors
          response to radiation
          response to mechanical stimulus
          response to organic substance
          response to ozone
          positive regulation of gene expression
          negative regulation of endopeptidase activity
          positive regulation of neuron projection development
          positive regulation of neuron maturation
          response to organic cyclic compound
          sensory perception of pain
          positive regulation of cell growth
          neuron projection development
          positive regulation of protein autophosphorylation
          response to lipopolysaccharide
          response to nicotine
          intracellular signal transduction
          nerve growth factor signaling pathway
          response to drug
          response to peptide hormone
          negative regulation of neuron apoptotic process
          regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of axon extension
          regulation of neurotransmitter secretion
          positive regulation of collateral sprouting
          neuron projection morphogenesis
          positive regulation of sequence-specific DNA binding transcription factor activity
          regulation of release of sequestered calcium ion into cytosol
          response to glucocorticoid
          positive regulation of neurotrophin TRK receptor signaling pathway
          neuron apoptotic process
          response to electrical stimulus
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of stem cell proliferation
          negative regulation of type B pancreatic cell apoptotic process
          molecular_function
          receptor signaling protein activity
          nerve growth factor receptor binding
          growth factor activity
          metalloendopeptidase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0