Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044243.1 | NP_001037708.1 | 10 | 957 | ||
XM_008762262.1 | XP_008760484.1 | 11 | 1009 | |||
GenBank mRNA | BC105618.1 | - | 10 | 957 | 7453153 |