Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107784.1 | NP_001101254.1 | 1 | 196 | ||
XM_008762269.1 | XP_008760491.1 | 1 | 227 |