Search
Search

Itch| Gene Symbol: Itch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005887.1 | NP_001005887.1 | 11 | 1235 | ||
| XM_008762335.1 | XP_008760557.1 | 13 | 1463 | |||
| XM_008762336.1 | XP_008760558.1 | 13 | 1388 | |||
| XM_008762337.1 | XP_008760559.1 | 10 | 1140 | |||
| GenBank mRNA | AY600518.1 | - | 11 | 1235 | ||