Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037979.1 | NP_001033068.1 | 6 | 970 | ||
XM_006237183.1 | XP_006237245.1 | 7 | 1276 | |||
XM_006237184.2 | XP_006237246.1 | 7 | 1147 | |||
XM_006237185.2 | XP_006237247.1 | 7 | 1162 | |||
XM_006237186.2 | XP_006237248.1 | 7 | 1291 | |||
GenBank mRNA | AY724514.1 | - | 7 | 1230 | ||
DQ148392.1 | - | 6 | 970 | |||
FQ232508.1 | - | 7 | 1249 |