Search
Search

PPFIA3| Gene Symbol: PPFIA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003660.3 | NP_003651.1 | 11 | 1619 | ||
| NR_103842.1 | 11 | 1619 | ||||
| XM_017027407.1 | XP_016882896.1 | 11 | 1619 | |||
| GenBank mRNA | AB014554.2 | - | 11 | 1602 | ||
| AK025972.1 | - | 8 | 819 | |||
| AK289757.1 | - | 11 | 1324 | |||
| AK293798.1 | - | 9 | 1070 | |||
| AK308483.1 | - | 9 | 1110 | |||
| BC101518.1 | - | 11 | 1345 | 8069024 | ||
| BC101520.1 | - | 11 | 1345 | 8069026 | ||
| BC143470.1 | - | 11 | 1345 | |||