Search
Search

Rmdn2| Gene Symbol: Rmdn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037200.1 | NP_001032277.1 | 2, 3 | 843 | ||
| XM_006239635.2 | XP_006239697.1 | 4, 5 | 1307 | |||
| XM_008764423.1 | XP_008762645.1 | 4, 5 | 1325 | |||
| GenBank mRNA | BC100261.1 | - | 2, 3 | 843 | 7368645 | |