Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MADD SIRNA Assays ›
Gene Symbol:
MADD
Gene Name:
MAP kinase activating death domain
Gene Aliases:
DENN, IG20, RAB3GEP
Chromosome Location:
Chr.11: 47269376 - 47330031 on Build GRCh38
UniGene ID:
Hs.82548
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16314
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MADD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135943.1
NP_001129415.1
28
4435
NM_001135944.1
NP_001129416.1
28
4402
NM_003682.3
NP_003673.3
31
4720
NM_130470.2
NP_569826.2
28
4709
NM_130471.2
NP_569827.2
29
4474
NM_130472.2
NP_569828.2
28
4414
NM_130473.2
NP_569829.2
29
4603
NM_130474.2
NP_569830.2
28
4414
NM_130475.2
NP_569831.1
31
4744
NM_130476.2
NP_569832.2
28
4564
XM_005253189.2
XP_005253246.1
31
4744
XM_005253196.2
XP_005253253.1
29
4627
XM_005253199.2
XP_005253256.1
28
4567
XM_005253200.2
XP_005253257.1
31
4741
XM_005253201.2
XP_005253258.1
29
4498
XM_005253203.2
XP_005253260.1
28
4438
XM_005253204.2
XP_005253261.1
28
4426
XM_005253205.1
XP_005253262.1
28
4438
XM_017018478.1
XP_016873967.1
31
4732
XM_017018479.1
XP_016873968.1
30
4678
XM_017018480.1
XP_016873969.1
29
4624
XM_017018481.1
XP_016873970.1
29
4618
XM_017018482.1
XP_016873971.1
29
4615
XM_017018483.1
XP_016873972.1
31
4603
XM_017018484.1
XP_016873973.1
28
4555
XM_017018485.1
XP_016873974.1
28
4516
XM_017018486.1
XP_016873975.1
30
4549
XM_017018487.1
XP_016873976.1
30
4543
XM_017018488.1
XP_016873977.1
31
4732
XM_017018489.1
XP_016873978.1
31
4693
XM_017018490.1
XP_016873979.1
30
4677
XM_017018491.1
XP_016873980.1
30
4643
XM_017018492.1
XP_016873981.1
30
4678
XM_017018493.1
XP_016873982.1
30
4659
XM_017018494.1
XP_016873983.1
29
4630
XM_017018495.1
XP_016873984.1
29
4627
XM_017018496.1
XP_016873985.1
29
4574
XM_017018497.1
XP_016873986.1
29
4579
XM_017018498.1
XP_016873987.1
31
4603
XM_017018499.1
XP_016873988.1
31
4572
XM_017018500.1
XP_016873989.1
28
4564
XM_017018501.1
XP_016873990.1
30
4517
XM_017018502.1
XP_016873991.1
30
4530
XM_017018503.1
XP_016873992.1
30
4514
XM_017018504.1
XP_016873993.1
30
4527
XM_017018505.1
XP_016873994.1
30
4505
XM_017018506.1
XP_016873995.1
30
4510
XM_017018507.1
XP_016873996.1
30
4530
XM_017018508.1
XP_016873997.1
29
4501
XM_017018509.1
XP_016873998.1
29
4454
XM_017018510.1
XP_016873999.1
29
4489
GenBank mRNA
AB002356.1
-
31
4716
AF440100.1
-
31
4694
AF440101.1
-
29
4577
AF440102.1
-
29
4448
AF440103.1
-
28
4388
AF440434.1
-
28
4388
AK292442.1
-
28
4366
AK308780.1
-
12
1172
BC040484.1
-
28
4419
5271798
U44953.1
-
28
4528
U77352.1
-
28
4677
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
cell surface receptor signaling pathway
regulation of tumor necrosis factor-mediated signaling pathway
regulation of Rab protein signal transduction
regulation of apoptotic process
positive regulation of GTPase activity
regulation of cell cycle
execution phase of apoptosis
regulation of extrinsic apoptotic signaling pathway via death domain receptors
regulation of extrinsic apoptotic signaling pathway
death receptor binding
protein binding
Rab guanyl-nucleotide exchange factor activity
protein kinase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0