Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240617.2 | XP_006240679.1 | 13 | 3462 | ||
XM_006240618.2 | XP_006240680.1 | 13 | 3166 | |||
XM_006240620.2 | XP_006240682.1 | 12 | 2878 | |||
XM_008764970.1 | XP_008763192.1 | 14 | 3229 | |||
XM_008764971.1 | XP_008763193.1 | 11 | 2905 | |||
XM_008764972.1 | XP_008763194.1 | 12 | 2848 |