Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbno2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108068.1 | NP_001101538.1 | 19 | 2284 | ||
XM_006240908.2 | XP_006240970.1 | 18 | 3566 | |||
XM_006240909.1 | XP_006240971.1 | 16 | 3027 | |||
XM_008765094.1 | XP_008763316.1 | 18 | 2149 | |||
XM_008765095.1 | XP_008763317.1 | 15 | 3712 |