Search
Search

Tbcel| Gene Symbol: Tbcel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014089.1 | NP_001014111.1 | 8 | 1254 | ||
| XM_006242859.2 | XP_006242921.1 | 8 | 3901 | |||
| XM_008766134.1 | XP_008764356.1 | 9 | 3864 | |||
| GenBank mRNA | BC087163.1 | - | 8 | 1254 | 7135124 | |