Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003728.3 | NP_003719.3 | 13 | 2552 | ||
XM_005263321.3 | XP_005263378.1 | 14 | 2609 | |||
XM_017008789.1 | XP_016864278.1 | 14 | 8969 | |||
XM_017008790.1 | XP_016864279.1 | 13 | 2197 | |||
GenBank mRNA | AF055634.1 | - | 13 | 2353 | ||
AK290500.1 | - | 13 | 2363 |