Search
Search

PTCH2| Gene Symbol: PTCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166292.1 | NP_001159764.1 | 9 | 1182 | ||
| NM_003738.4 | NP_003729.3 | 9 | 1182 | |||
| GenBank mRNA | AF087651.1 | - | 9 | 1170 | ||
| AF091501.1 | - | 9 | 1170 | |||
| AF119569.1 | - | 9 | 1182 | |||
| AY358555.1 | - | 9 | 1170 | |||
| AY359016.1 | - | 9 | 1174 | |||
| BC143712.1 | - | 8 | 1042 | |||