Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AKR1C3 SIRNA Assays ›
          Gene Symbol:
          AKR1C3
          Gene Name:
          aldo-keto reductase family 1, member C3
          Gene Aliases:
          DD3, DDX, HA1753, HAKRB, HAKRe, HSD17B5, PGFS, hluPGFS
          Chromosome Location:
          Chr.10: 5048766 - 5107686 on Build GRCh38
          UniGene ID:
          Hs.78183
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s16448

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AKR1C3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001253908.1 NP_001240837.1 6 645
          NM_003739.5 NP_003730.4 6 668
          GenBank mRNA AB018580.1 - 6 646
          AF149416.2 - 6 610
          AK290365.1 - 6 628
          AK296829.1 - 6 712
          AK315411.1 - 6 628
          BC001479.2 - 6 655 2988160
          BC019230.1 - 6 625 3682448
          BT007286.1 - 6 599
          D17793.1 - 6 650
          DQ269985.1 - 7 599
          S68288.1 - 6 609

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          retinoid metabolic process
          prostaglandin metabolic process
          G-protein coupled receptor signaling pathway
          response to nutrient
          steroid metabolic process
          positive regulation of cell proliferation
          male gonad development
          cellular response to starvation
          positive regulation of cell death
          farnesol catabolic process
          cyclooxygenase pathway
          keratinocyte differentiation
          cellular response to reactive oxygen species
          progesterone metabolic process
          retinol metabolic process
          retinal metabolic process
          multicellular organismal macromolecule metabolic process
          daunorubicin metabolic process
          doxorubicin metabolic process
          regulation of retinoic acid receptor signaling pathway
          positive regulation of protein kinase B signaling
          oxidation-reduction process
          testosterone biosynthetic process
          renal absorption
          cellular response to cadmium ion
          cellular response to calcium ion
          cellular response to prostaglandin stimulus
          cellular response to corticosteroid stimulus
          cellular response to jasmonic acid stimulus
          cellular response to prostaglandin D stimulus
          negative regulation of retinoic acid biosynthetic process
          regulation of testosterone biosynthetic process
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          retinal dehydrogenase activity
          alditol:NADP+ 1-oxidoreductase activity
          aldo-keto reductase (NADP) activity
          retinol dehydrogenase activity
          oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
          phenanthrene 9,10-monooxygenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          prostaglandin H2 endoperoxidase reductase activity
          prostaglandin D2 11-ketoreductase activity
          geranylgeranyl reductase activity
          ketoreductase activity
          prostaglandin-F synthase activity
          15-hydroxyprostaglandin-D dehydrogenase (NADP+) activity
          androsterone dehydrogenase activity
          testosterone dehydrogenase (NAD+) activity
          testosterone 17-beta-dehydrogenase (NADP+) activity
          ketosteroid monooxygenase activity
          trans-1,2-dihydrobenzene-1,2-diol dehydrogenase activity
          indanol dehydrogenase activity
          delta4-3-oxosteroid 5beta-reductase activity
          NADP-retinol dehydrogenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0